missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GB 2763 Organochlorine Pesticides Mixture 110 20-100 μg/mL in Toluene:n-Hexane, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | 2,4'-DDT∣4,4'-DDD∣4,4'-DDE∣4,4'-DDT∣Aldrin∣alpha-Endosulfan∣alpha-HCH∣beta-Endosulfan∣beta-HCH∣Cis-Chlordane (Alpha Isomer)∣delta-HCH∣Dieldrin∣Endosulfan-sulfate∣gamma-HCH∣Heptachlor∣Heptachlor-endo-epoxide (trans-, isomer A)∣Heptachlor-exo-epoxide (cis-, isomer B)∣oxy-Chlordane∣Trans-Chlordane (Gamma Isomer) |
| CAS | 789-02-6, 72-54-8, 72-55-9, 50-29-3, 309-00-2, 959-98-8 |
| Analyte or Component Names | 2,4'-DDT∣4,4'-DDD∣4,4'-DDE∣4,4'-DDT∣Aldrin∣alpha-Endosulfan∣alpha-HCH∣beta-Endosulfan∣beta-HCH∣Cis-Chlordane (Alpha Isomer)∣delta-HCH∣Dieldrin∣Endosulfan-sulfate∣gamma-HCH∣Heptachlor∣Heptachlor-endo-epoxide (trans-, isomer A)∣Heptachlor-exo-epoxide (cis-, isomer B)∣oxy-Chlordane∣Trans-Chlordane (Gamma Isomer) |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | n-Hexane/Toluene |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | Cis-Chlordane (Alpha Isomer) 20 μg/mL∣oxy-Chlordane 20 μg/mL∣Trans-Chlordane (Gamma Isomer) 20 μg/mL∣Heptachlor 50 μg/mL∣Heptachlor-endo-epoxide (trans-, isomer A) 50 μg/mL∣Heptachlor-exo-epoxide (cis-, isomer B) 50 μg/mL∣2,4'-DDT 100 μg/mL∣4,4'-DDD 100 μg/mL∣4,4'-DDE 100 μg/mL∣4,4'-DDT 100 μg/mL∣Aldrin 100 μg/mL∣alpha-Endosulfan 100 μg/mL∣alpha-HCH 100 μg/mL∣beta-Endosulfan 100 μg/mL∣beta-HCH 100 μg/mL∣delta-HCH 100 μg/mL∣Dieldrin 100 μg/mL∣Endosulfan-sulfate 100 μg/mL∣gamma-HCH 100 μg/mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction