missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GB 23200.121-2021 Pesticide Mixture 9 50 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Avermectin B1a∣Bifenox∣Bromuconazole∣Chlordimeform free base∣Chlorfenvinphos∣Clethodim Sulfoxide∣Clofentezine∣Cyflumetofen∣Ethofumesate∣Fenarimol∣Fenazaquin∣Fenbuconazole∣Fenoxaprop-ethyl∣Fenpropidin∣Fenpropimorph∣Fipronil-desulfinyl∣Fluazinam∣Flucythrinate∣Fludioxonil∣Flumetralin∣Flusilazole∣Flutriafol∣Hexaconazole∣Imibenconazole∣Ipconazole∣Isoxaflutole-diketonitrile∣Metazachlor∣Metconazole∣Metrafenone∣Oxyfluorfen∣Paclobutrazol∣Penconazole∣Phenamacril∣Propiconazole∣Pyrametostrobin∣Pyridalyl∣Pyrisoxazole∣Tetraconazole∣Triadimefon∣Trichlorphon∣Tricyclazole∣Trifloxystrobin∣Triflumizole∣Triticonazole∣Uniconazole |
| CAS | 65195-55-3, 42576-02-3, 116255-48-2, 6164-98-3, 470-90-6, 111031-14-2 |
| Analyte or Component Names | Avermectin B1a∣Bifenox∣Bromuconazole∣Chlordimeform free base∣Chlorfenvinphos∣Clethodim Sulfoxide∣Clofentezine∣Cyflumetofen∣Ethofumesate∣Fenarimol∣Fenazaquin∣Fenbuconazole∣Fenoxaprop-ethyl∣Fenpropidin∣Fenpropimorph∣Fipronil-desulfinyl∣Fluazinam∣Flucythrinate∣Fludioxonil∣Flumetralin∣Flusilazole∣Flutriafol∣Hexaconazole∣Imibenconazole∣Ipconazole∣Isoxaflutole-diketonitrile∣Metazachlor∣Metconazole∣Metrafenone∣Oxyfluorfen∣Paclobutrazol∣Penconazole∣Phenamacril∣Propiconazole∣Pyrametostrobin∣Pyridalyl∣Pyrisoxazole∣Tetraconazole∣Triadimefon∣Trichlorphon∣Tricyclazole∣Trifloxystrobin∣Triflumizole∣Triticonazole∣Uniconazole |
| Shipping Condition | +20°C |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Acetonitrile |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | Avermectin B1a 50 μg/mL∣Bifenox 50 μg/mL∣Bromuconazole 50 μg/mL∣Chlordimeform free base 50 μg/mL∣Chlorfenvinphos 50 μg/mL∣Clethodim Sulfoxide 50 μg/mL∣Clofentezine 50 μg/mL∣Cyflumetofen 50 μg/mL∣Ethofumesate 50 μg/mL∣Fenarimol 50 μg/mL∣Fenazaquin 50 μg/mL∣Fenbuconazole 50 μg/mL∣Fenoxaprop-ethyl 50 μg/mL∣Fenpropidin 50 μg/mL∣Fenpropimorph 50 μg/mL∣Fipronil-desulfinyl 50 μg/mL∣Fluazinam 50 μg/mL∣Flucythrinate 50 μg/mL∣Fludioxonil 50 μg/mL∣Flumetralin 50 μg/mL∣Flusilazole 50 μg/mL∣Flutriafol 50 μg/mL∣Hexaconazole 50 μg/mL∣Imibenconazole 50 μg/mL∣Ipconazole 50 μg/mL∣Isoxaflutole-diketonitrile 50 μg/mL∣Metazachlor 50 μg/mL∣Metconazole 50 μg/mL∣Metrafenone 50 μg/mL∣Oxyfluorfen 50 μg/mL∣Paclobutrazol 50 μg/mL∣Penconazole 50 μg/mL∣Phenamacril 50 μg/mL∣Propiconazole 50 μg/mL∣Pyrametostrobin 50 μg/mL∣Pyridalyl 50 μg/mL∣Pyrisoxazole 50 μg/mL∣Tetraconazole 50 μg/mL∣Triadimefon 50 μg/mL∣Trichlorphon 50 μg/mL∣Tricyclazole 50 μg/mL∣Trifloxystrobin 50 μg/mL∣Triflumizole 50 μg/mL∣Triticonazole 50 μg/mL∣Uniconazole 50 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?