missing translation for 'onlineSavingsMsg'
Learn More

GB 23200.121-2021 Pesticide Mixture 8 50 μg/mL in Acetonitrile:Methanol, Dr. Ehrenstorfer

Product Code. 30325625
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30325625 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30325625 Supplier Dr. Ehrenstorfer Supplier No. DREA50000728MC
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (±)-Fenpropathrin∣(E)-Fenpyroximate∣Azoxystrobin∣Bifenthrin∣Bioresmethrin∣Bitertanol∣Coumoxystrobin∣Cycloxydim∣Cyproconazole∣Deltamethrin∣Diclofop methyl∣Difenoconazole∣Diniconazole∣Dinocap∣Enoxastrobin∣Epoxiconazole∣Etofenprox∣Etoxazole∣Fenvalerate∣Fluazifop-butyl∣Fluoroglycofen-ethyl∣Isoprothiolane∣Ivermectin∣Kresoxim-methyl∣Lactofen∣Methoprene∣Myclobutanil∣Oxadiazon∣Permethrin∣Picoxystrobin∣Piperonyl butoxide∣Propaquizafop∣Propargite∣Pyraflufen-ethyl∣Pyrethrin 1∣Pyriftalid∣Pyriproxyfen∣Quizalofop-ethyl∣Rotenone∣Spinetoram∣Spirodiclofen∣Spiromesifen∣Tebuconazole∣Thiacloprid∣Thiamethoxam∣Triadimenol
CAS 39515-41-8, 134098-61-6, 131860-33-8, 82657-04-3, 28434-01-7, 55179-31-2
Analyte or Component Names (±)-Fenpropathrin∣(E)-Fenpyroximate∣Azoxystrobin∣Bifenthrin∣Bioresmethrin∣Bitertanol∣Coumoxystrobin∣Cycloxydim∣Cyproconazole∣Deltamethrin∣Diclofop methyl∣Difenoconazole∣Diniconazole∣Dinocap∣Enoxastrobin∣Epoxiconazole∣Etofenprox∣Etoxazole∣Fenvalerate∣Fluazifop-butyl∣Fluoroglycofen-ethyl∣Isoprothiolane∣Ivermectin∣Kresoxim-methyl∣Lactofen∣Methoprene∣Myclobutanil∣Oxadiazon∣Permethrin∣Picoxystrobin∣Piperonyl butoxide∣Propaquizafop∣Propargite∣Pyraflufen-ethyl∣Pyrethrin 1∣Pyriftalid∣Pyriproxyfen∣Quizalofop-ethyl∣Rotenone∣Spinetoram∣Spirodiclofen∣Spiromesifen∣Tebuconazole∣Thiacloprid∣Thiamethoxam∣Triadimenol
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Solvent or Matrix Acetonitrile/Methanol
Quantity 1 mL
Concentration or Composition (by Analyte or Components) (±)-Fenpropathrin 50 μg/mL∣(E)-Fenpyroximate 50 μg/mL∣Azoxystrobin 50 μg/mL∣Bifenthrin 50 μg/mL∣Bioresmethrin 50 μg/mL∣Bitertanol 50 μg/mL∣Coumoxystrobin 50 μg/mL∣Cycloxydim 50 μg/mL∣Cyproconazole 50 μg/mL∣Deltamethrin 50 μg/mL∣Diclofop methyl 50 μg/mL∣Difenoconazole 50 μg/mL∣Diniconazole 50 μg/mL∣Dinocap 50 μg/mL∣Enoxastrobin 50 μg/mL∣Epoxiconazole 50 μg/mL∣Etofenprox 50 μg/mL∣Etoxazole 50 μg/mL∣Fenvalerate 50 μg/mL∣Fluazifop-butyl 50 μg/mL∣Fluoroglycofen-ethyl 50 μg/mL∣Isoprothiolane 50 μg/mL∣Ivermectin 50 μg/mL∣Kresoxim-methyl 50 μg/mL∣Lactofen 50 μg/mL∣Methoprene 50 μg/mL∣Myclobutanil 50 μg/mL∣Oxadiazon 50 μg/mL∣Permethrin 50 μg/mL∣Picoxystrobin 50 μg/mL∣Piperonyl butoxide 50 μg/mL∣Propaquizafop 50 μg/mL∣Propargite 50 μg/mL∣Pyraflufen-ethyl 50 μg/mL∣Pyrethrin 1 50 μg/mL∣Pyriftalid 50 μg/mL∣Pyriproxyfen 50 μg/mL∣Quizalofop-ethyl 50 μg/mL∣Rotenone 50 μg/mL∣Spinetoram 50 μg/mL∣Spirodiclofen 50 μg/mL∣Spiromesifen 50 μg/mL∣Tebuconazole 50 μg/mL∣Thiacloprid 50 μg/mL∣Thiamethoxam 50 μg/mL∣Triadimenol 50 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.