missing translation for 'onlineSavingsMsg'
Learn More

GB 23200.121-2021 Pesticide Mixture 7 50 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30313961
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30313961 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30313961 Supplier Dr. Ehrenstorfer Supplier No. DREA50000727AL
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (E)-Nitenpyram∣2-Diethylaminoethyl Hexanoate∣Acetamiprid∣Ametoctradin∣Ametryn∣Atrazine∣Bupirimate∣Butralin∣Clothianidin∣Cyanazine∣Cyprodinil∣Diclobutrazol∣Dinotefuran∣Fipronil Sulfide∣Flurtamone∣Imidacloprid∣Imidaclothiz∣Metribuzin∣Pendimethalin∣Prometryn∣Pyraoxystrobin∣Pyrimethanil∣Simazine∣Simetryn∣Spinosyn A∣Spinosyn D∣tau-Fluvalinate∣Terbuthylazine∣Thiabendazole∣Thifluzamide∣Triflumizole-amino
CAS 150824-47-8, 10369-83-2, 135410-20-7, 865318-97-4, 834-12-8, 1912-24-9
Analyte or Component Names (E)-Nitenpyram∣2-Diethylaminoethyl Hexanoate∣Acetamiprid∣Ametoctradin∣Ametryn∣Atrazine∣Bupirimate∣Butralin∣Clothianidin∣Cyanazine∣Cyprodinil∣Diclobutrazol∣Dinotefuran∣Fipronil Sulfide∣Flurtamone∣Imidacloprid∣Imidaclothiz∣Metribuzin∣Pendimethalin∣Prometryn∣Pyraoxystrobin∣Pyrimethanil∣Simazine∣Simetryn∣Spinosyn A∣Spinosyn D∣tau-Fluvalinate∣Terbuthylazine∣Thiabendazole∣Thifluzamide∣Triflumizole-amino
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Solvent or Matrix Acetonitrile
Quantity 1 mL
Concentration or Composition (by Analyte or Components) (E)-Nitenpyram 50 μg/mL∣2-Diethylaminoethyl Hexanoate 50 μg/mL∣Acetamiprid 50 μg/mL∣Ametoctradin 50 μg/mL∣Ametryn 50 μg/mL∣Atrazine 50 μg/mL∣Bupirimate 50 μg/mL∣Butralin 50 μg/mL∣Clothianidin 50 μg/mL∣Cyanazine 50 μg/mL∣Cyprodinil 50 μg/mL∣Diclobutrazol 50 μg/mL∣Dinotefuran 50 μg/mL∣Fipronil Sulfide 50 μg/mL∣Flurtamone 50 μg/mL∣Imidacloprid 50 μg/mL∣Imidaclothiz 50 μg/mL∣Metribuzin 50 μg/mL∣Pendimethalin 50 μg/mL∣Prometryn 50 μg/mL∣Pyraoxystrobin 50 μg/mL∣Pyrimethanil 50 μg/mL∣Simazine 50 μg/mL∣Simetryn 50 μg/mL∣Spinosyn A 50 μg/mL∣Spinosyn D 50 μg/mL∣tau-Fluvalinate 50 μg/mL∣Terbuthylazine 50 μg/mL∣Thiabendazole 50 μg/mL∣Thifluzamide 50 μg/mL∣Triflumizole-amino 50 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.