missing translation for 'onlineSavingsMsg'
Learn More

GB 23200.121-2021 Pesticide Mixture 2 50 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30327674
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30327674 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30327674 Supplier Dr. Ehrenstorfer Supplier No. DREA50000722AL
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Amidosulfuron∣Bensulfuron-methyl∣Chlorsulfuron∣Cinosulfuron∣Ethoxysulfuron∣Etrimfos∣Fenpyrazamine∣Flucetosulfuron∣Halosulfuron-methyl∣Iodosulfuron-methyl sodium∣Mesosulfuron-methyl∣Metazosulfuron∣Metsulfuron-methyl∣Propyrisulfuron∣Pyrazosulfuron-ethyl∣Thifensulfuron-methyl∣Triasulfuron∣Tribenuron-methyl (technical)∣Triflusulfuron-methyl∣Tritosulfuron
CAS 120923-37-7, 83055-99-6, 64902-72-3, 94593-91-6, 126801-58-9, 38260-54-7
Analyte or Component Names Amidosulfuron∣Bensulfuron-methyl∣Chlorsulfuron∣Cinosulfuron∣Ethoxysulfuron∣Etrimfos∣Fenpyrazamine∣Flucetosulfuron∣Halosulfuron-methyl∣Iodosulfuron-methyl sodium∣Mesosulfuron-methyl∣Metazosulfuron∣Metsulfuron-methyl∣Propyrisulfuron∣Pyrazosulfuron-ethyl∣Thifensulfuron-methyl∣Triasulfuron∣Tribenuron-methyl (technical)∣Triflusulfuron-methyl∣Tritosulfuron
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Solvent or Matrix Acetonitrile
Quantity 1 mL
Concentration or Composition (by Analyte or Components) Amidosulfuron 50 μg/mL∣Bensulfuron-methyl 50 μg/mL∣Chlorsulfuron 50 μg/mL∣Cinosulfuron 50 μg/mL∣Ethoxysulfuron 50 μg/mL∣Etrimfos 50 μg/mL∣Fenpyrazamine 50 μg/mL∣Flucetosulfuron 50 μg/mL∣Halosulfuron-methyl 50 μg/mL∣Iodosulfuron-methyl sodium 50 μg/mL∣Mesosulfuron-methyl 50 μg/mL∣Metazosulfuron 50 μg/mL∣Metsulfuron-methyl 50 μg/mL∣Propyrisulfuron 50 μg/mL∣Pyrazosulfuron-ethyl 50 μg/mL∣Thifensulfuron-methyl 50 μg/mL∣Triasulfuron 50 μg/mL∣Tribenuron-methyl (technical) 50 μg/mL∣Triflusulfuron-methyl 50 μg/mL∣Tritosulfuron 50 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.