missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GB 23200.121-2021 Pesticide Mixture 1 50 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Aldicarb∣Aldicarb-sulfone∣Aldicarb-sulfoxide∣Benazolin-ethyl ester∣Bendiocarb∣Carbaryl∣Carbofuran∣Carbofuran-3-hydroxy∣Chlorantraniliprole∣Chlorpropham∣Diethofencarb∣Famoxadone∣Fenobucarb∣Fenothiocarb∣Fenoxycarb∣Iprovalicarb∣Isoprocarb∣Methiocarb∣Methiocarb sulfone∣Methiocarb sulfoxide∣Methomyl∣Metolcarb∣Oxadiargyl∣Oxadixyl∣Oxamyl∣Phenmedipham∣Phosalone∣Pirimicarb∣Pirimicarb-desmethyl∣Pirimicarb-desmethyl-formamido∣Promecarb∣Propamocarb free base∣Propoxur∣Prosulfocarb∣Pyraclostrobin∣Pyridaphenthion∣Triallate |
| CAS | 116-06-3, 1646-88-4, 1646-87-3, 25059-80-7, 22781-23-3, 63-25-2 |
| Analyte or Component Names | Aldicarb∣Aldicarb-sulfone∣Aldicarb-sulfoxide∣Benazolin-ethyl ester∣Bendiocarb∣Carbaryl∣Carbofuran∣Carbofuran-3-hydroxy∣Chlorantraniliprole∣Chlorpropham∣Diethofencarb∣Famoxadone∣Fenobucarb∣Fenothiocarb∣Fenoxycarb∣Iprovalicarb∣Isoprocarb∣Methiocarb∣Methiocarb sulfone∣Methiocarb sulfoxide∣Methomyl∣Metolcarb∣Oxadiargyl∣Oxadixyl∣Oxamyl∣Phenmedipham∣Phosalone∣Pirimicarb∣Pirimicarb-desmethyl∣Pirimicarb-desmethyl-formamido∣Promecarb∣Propamocarb free base∣Propoxur∣Prosulfocarb∣Pyraclostrobin∣Pyridaphenthion∣Triallate |
| Shipping Condition | +20°C |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Acetonitrile |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | Aldicarb 50 μg/mL∣Aldicarb-sulfone 50 μg/mL∣Aldicarb-sulfoxide 50 μg/mL∣Benazolin-ethyl ester 50 μg/mL∣Bendiocarb 50 μg/mL∣Carbaryl 50 μg/mL∣Carbofuran 50 μg/mL∣Carbofuran-3-hydroxy 50 μg/mL∣Chlorantraniliprole 50 μg/mL∣Chlorpropham 50 μg/mL∣Diethofencarb 50 μg/mL∣Famoxadone 50 μg/mL∣Fenobucarb 50 μg/mL∣Fenothiocarb 50 μg/mL∣Fenoxycarb 50 μg/mL∣Iprovalicarb 50 μg/mL∣Isoprocarb 50 μg/mL∣Methiocarb 50 μg/mL∣Methiocarb sulfone 50 μg/mL∣Methiocarb sulfoxide 50 μg/mL∣Methomyl 50 μg/mL∣Metolcarb 50 μg/mL∣Oxadiargyl 50 μg/mL∣Oxadixyl 50 μg/mL∣Oxamyl 50 μg/mL∣Phenmedipham 50 μg/mL∣Phosalone 50 μg/mL∣Pirimicarb 50 μg/mL∣Pirimicarb-desmethyl 50 μg/mL∣Pirimicarb-desmethyl-formamido 50 μg/mL∣Promecarb 50 μg/mL∣Propamocarb free base 50 μg/mL∣Propoxur 50 μg/mL∣Prosulfocarb 50 μg/mL∣Pyraclostrobin 50 μg/mL∣Pyridaphenthion 50 μg/mL∣Triallate 50 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?