Learn More
BRAND™ Gay-Lussac Pycnometer
Product Code. 16305145 Shop All Brand Products
Change view
Click to view available options
Gay-Lussac Pycnometer
Product Code.
16305145 Gay-Lussac Pycnometer
1 options
Product Code. 16305145 Supplier BRAND™ Supplier No. DAKKS/43308
Pycnometer made of borosilicate glass 3.3 and calibrated.
  • Material: Borosilicate glass 3.3
  • Type: Gay-Lussac
  • DIN ISO 3507
  • Calibrated to contain (TC, In)
  • Includes individual certificate
  • Stopper NS 10/19 with capillary, top of stopper ground and polished
TRUSTED_SUSTAINABILITY
Specifications
Calibration To Contain
Capacity (Metric) 10 mL
Material Borosilicate Glass (3.3)
Shape Gay-Lussac
Autoclavable Autoclavable
Disposable Reusable
Graduated Yes
Mouth Style Narrow
Sterility Non-sterile
Bottom Shape Flat