missing translation for 'onlineSavingsMsg'
Learn More

Gavestinel, TRC

Product Code. 30398000
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30398000 10 mg 10mg
30399884 5 mg -
30392007 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30398000 Supplier Toronto Research Chemicals Supplier No. TRCG59661810MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 153436-38-5
Molecular Formula C18H11Cl2N2NaO3
Molecular Weight (g/mol) 397.19
Synonym (E)-4,6-Dichloro-3-[3-oxo-3-(phenylamino)-1-propenyl]-1H-indole-2-carboxylic Acid Monosodium Salt, GV 150526A, 4,6-Dichloro-3-[(1E)-3-oxo-3-(phenylamino)-1-propenyl]-1H-indole-2-carboxylic Acid Monosodium Salt
IUPAC Name sodium;3-[(E)-3-anilino-3-oxoprop-1-enyl]-4,6-dichloro-1H-indole-2-carboxylate
SMILES [Na+].[O-]C(=O)c1[nH]c2cc(Cl)cc(Cl)c2c1\C=C\C(=O)Nc3ccccc3

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C18H12Cl2N2O3.Na/c19-10-8-13(20)16-12(17(18(24)25)22-14(16)9-10)6-7-15(23)21-11-4-2-1-3-5-11;/h1-9,22H,(H,21,23)(H,24,25);/q;+1/p-1/b7-6+;
Formula Weight 396.0044
Chemical Name or Material Gavestinel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.