missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Gavestinel
GREENER_CHOICE

Product Code. 18065772
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18065772 50 mg 50mg
18045492 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18065772 Supplier Tocris Bioscience™ Supplier No. 2348/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective glycine site antagonist; orally available and active in vivo

Highly potent and selective non-competitive antagonist acting at the strychnine-insensitive glycine binding site of the NMDA receptor-channel complex (Kd = 0.8 nM). Displays > 1000-fold selectivity over NMDA, AMPA and kainate binding sites. Orally bioavailable and active in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 153436-38-5
Molecular Formula C18H12Cl2N2NaO3

Specifications

Quantity 50 mg
Formula Weight 397.19
Purity >98%
Molecular Weight (g/mol) 398.195
Chemical Name or Material Gavestinel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.