missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GATSL2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA572114
This item is not returnable.
View return policy
Description
GATSL2 Polyclonal Antibody for Western Blot, IHC (P)
Functions as a negative regulator of the TORC1 signaling pathway through the GATOR complex. As part of homodimers or heterodimers with CASTOR1, directly binds and inhibits the GATOR subcomplex GATOR2 and thereby mTORC1. Does not directly bind arginine, but binding of arginine to CASTOR1 disrupts the interaction of CASTOR2-containing heterodimers with GATOR2 which can in turn activate mTORC1 and the TORC1 signaling pathway.
Specifications
| GATSL2 | |
| Polyclonal | |
| Unconjugated | |
| CASTOR2 | |
| CASTOR2; Cellular arginine sensor for mTORC1 protein 2; Cytosolic arginine sensor for mTORC1 subunit 2; GATL2; GATS protein like 2; GATS protein-like 1; GATS protein-like 2; GATSL1; GATSL2; GATS-like protein 1; GATS-like protein 2 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 729438 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| A6NHX0 | |
| CASTOR2 | |
| KLH conjugated synthetic peptide between 25-51 amino acids from the N-terminal region of human GATSL2. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction