missing translation for 'onlineSavingsMsg'
Learn More

Gatifloxacin sesquihydrate

Product Code. 15497229
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 15497229

Brand: Thermo Scientific Alfa Aesar J66036.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A broad spectrum antibiotic | CAS: 180200-66-2 | C19H22FN3O4 | 375.40 g/mol

A broad spectrum antibiotic

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 180200-66-2
Molecular Formula C19H22FN3O4
Molecular Weight (g/mol) 375.40
MDL Number MFCD00895399
InChI Key XUBOMFCQGDBHNK-UHFFFAOYNA-N
PubChem CID 5379
ChEBI CHEBI:5280
IUPAC Name 1-cyclopropyl-6-fluoro-8-methoxy-7-(3-methylpiperazin-1-yl)-4-oxoquinoline-3-carboxylic acid
SMILES COC1=C2N(C=C(C(O)=O)C(=O)C2=CC(F)=C1N1CCNC(C)C1)C1CC1

Specifications

Type Gatifloxacin Sesquihydrate
Physical Form Powder
Color White to Yellow
Quantity 1 g
Boiling Point 684°C
Merck Index 14,4376
Solubility Information Soluble in 1N NaOH at 10mg/mL
Formula Weight 402.42
Chemical Name or Material Gatifloxacin sesquihydrate
compliance-icons
Product Identifier
  • Gatifloxacin sesquihydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Gatifloxacin sesquihydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.