missing translation for 'onlineSavingsMsg'
Learn More

Gatifloxacin Sesquihydrate, Mikromol

Product Code. 30461519
Click to view available options
Quantity:
250 mg
This item is not returnable. View return policy

Product Code. 30461519

Brand: Mikromol MM0884.000250

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Gatifloxacin Sesquihydrate, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Gatifloxacin Sesquihydrate
CAS 180200-66-2
Analyte or Component Names Gatifloxacin Sesquihydrate
Shipping Condition Room Temperature
Molecular Formula 2 C19 H22 F N3 O4 . 3 H2 O
Certifications/Compliance ISO 17034
Synonym 1-Cyclopropyl-6-fluoro-1,4-dihydro-8-methoxy-7-(3-methyl-1-piperazinyl)-4-oxo-3-quinolinecarboxylic acid Hydrate (2:3), (+/-)-1-Cyclopropyl-6-fluoro-1,4-dihydro-8-methoxy-7-(3-methyl-1-piperazinyl)-4-oxo-3-quinolinecarboxylic acid Sesquihydrate, Gatifloxa
InChI Formula InChI=1S/2C19H22FN3O4.3H2O/c2*1-10-8-22(6-5-21-10)16-14(20)7-12-15(18(16)27-2)23(11-3-4-11)9-13(17(12)24)19(25)26;;;/h2*7,9-11,21H,3-6,8H2,1-2H3,(H,25,26);3*1H2
Solution Type Neat
SMILES O.O.O.COc1c(N2CCNC(C)C2)c(F)cc3C(=O)C(=CN(C4CC4)c13)C(=O)O.COc5c(N6CCNC(C)C6)c(F)cc7C(=O)C(=CN(C8CC8)c57)C(=O)O
IUPAC Name 1-cyclopropyl-6-fluoro-8-methoxy-7-(3-methylpiperazin-1-yl)-4-oxoquinoline-3-carboxylic acid;trihydrate
Molecular Weight (g/mol) 804.834
Quantity 250 mg
Formula Weight 804.351 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.