missing translation for 'onlineSavingsMsg'
Learn More

Gatifloxacin, MedChemExpress

Product Code. p-200098107
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 16471059

Brand: Medchem Express HY10581

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Gatifloxacin (AM-1155; BMS-206584; PD135432) is a potent fluoroquinolone antibiotic with broad-spectrum antibacterial activity. Gatifloxacin inhibits bacterial type II topoisomerases (IC50=13.8 μg/ml for S. aureus topoisomerase IV) and E. coli DNA gyrase (IC50=0.109 μg/ml). Gatifloxacin can be used to treat bacterial conjunctivitis in vivo.

Gatifloxacin (AM-1155

Specifications

Chemical Name or Material Gatifloxacin
Physical Form Solid
CAS 112811-59-3
Color White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C19H22FN3O4
For Use With (Application) COVID-19-anti-virus
Synonym AM-1155 BMS-206584 PD135432
Solubility Information DMSO : 2 mg/mL (5.33 mM; Need ultrasonic) ∣H2O : 1 mg/mL (2.66 mM; Need ultrasonic)
SMILES CC1NCCN(C2=C(F)C=C3C(N(C4CC4)C=C(C(O)=O)C3=O)=C2OC)C1
Molecular Weight (g/mol) 375.39
Formula Weight 375.39
Percent Purity 99.37%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.