missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gatifloxacin hydrochloride, MedChemExpress
Description
Gatifloxacin hydrochloride (AM-1155
Specifications
Specifications
| Chemical Name or Material | Gatifloxacin hydrochloride |
| Physical Form | Solid |
| CAS | 121577-32-0 |
| Color | White |
| Quantity | 1 g |
| Purity Grade Notes | Research |
| Molecular Formula | C19H23ClFN3O4 |
| For Use With (Application) | COVID-19-anti-virus |
| Synonym | AM-1155 hydrochlorideBMS-206584 hydrochloridePD135432 hydrochloride |
| Solubility Information | H2O : 10 mg/mL (24.28 mM; Need ultrasonic) ∣DMSO : 5 mg/mL (12.14 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?