Learn More
Specac™ Gateway™ ATR Water Heated/Cooled Trough Top Plate With ZnSe Crystal
Product Code. 15250138
Encompass_Preferred
Change view
Click to view available options
For Use With (Equipment):
Gateway ATR System GS11165
Product Code. For Use With (Equipment)
15250138 Gateway ATR System GS11165
1 options
Product Code. 15250138 Supplier Specac™ Supplier No. GS11139
Estimated Shipment: 22-09-2026
Add to basket
Add to basket
Water Heated/Cooled Top-Plate Assembly for use in the Gateway ATR system GS11165. Can work sub ambient to 90°C by thermocirculating fluid.
Specifications
For Use With (Equipment) Gateway ATR System GS11165