Learn More
Specac™ Gateway™ ATR Electrically Heated Trough Top Plate With ZnSe Crystal & ATC
Product Code. 15210148
Encompass_Preferred
Click to view available options
For Use With (Equipment):
Gateway ATR Accessory GS11165
Product Code. 15210148
Brand: Specac™ GS11155
Electrically Heated trough Top-Plate with ZnSe crystal for used in the Gateway ATR Accessory GS11165. This Top-Plate is heatable to 200°C.
Specifications
For Use With (Equipment) Gateway ATR Accessory GS11165