Learn More
Specac™ Gateway™ ATR 550μl Thermostabilised Flow Through Top-Plate Assembly
Product Code. 15290128
Encompass_Preferred
Change view
Click to view available options
For Use With (Equipment):
Gateway ATR System GS11165
Product Code. For Use With (Equipment)
15290128 Gateway ATR System GS11165
1 options
Product Code. 15290128 Supplier Specac™ Supplier No. GS11118
Estimated Shipment: 04-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Heatable Top-Plate for use on the Gateway ATR system GS11165. This is a 550 microliter thermostabilised assembly with a ZnSe crystal and heatable to 90°C.
Specifications
For Use With (Equipment) Gateway ATR System GS11165