Learn More
Specac™ Gateway™ ATR 550μl Flow Through Top-Plate Assembly
Product Code. 15280128
Encompass_Preferred
Click to view available options
For Use With (Equipment):
Gateway ATR System GS11165
Product Code. 15280128
Brand: Specac™ GS11116
Room temperature flowthrough Top-Plate with ZnSe crystal for use on the Gateway ATR system GS11165. Volume of 550μL.
Specifications
For Use With (Equipment) Gateway ATR System GS11165
Volume (Metric) 550 μL