missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GATC Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA589654
This item is not returnable.
View return policy
Description
Immunogen sequence: MWSRLVWLGL RAPLGGRQGF TSKADPQGSG RITAAVIEHL ERLALVDFGS REAVARLEKA IAFADRLRAV DTDGVEPMES VLEDRCLYLR SDNVVEGNCA DELLQNSHRV VEEYFVAPPG NISLPKLDEQ EPFPHS; Positive Samples: MCF7; Cellular Location: Mitochondrion.
GATC (glutamyl-tRNA(Gln) amidotransferase, subunit C) is a protein-coding gene. GO annotations related to this gene include glutaminyl-tRNA synthase (glutamine-hydrolyzing) activity. An important paralog of this gene is ENSG00000111780.
Specifications
| GATC | |
| Polyclonal | |
| Unconjugated | |
| GATC | |
| 15E1.2; 2010003O18Rik; Gatc; gatC-like protein; glu-AdT subunit C; glu-AdT subunit C {ECO:0000255; glutamyl-tRNA amidotransferase subunit C; glutamyl-tRNA(Gln) amidotransferase subunit C, mitochondrial; glutamyl-tRNA(Gln) amidotransferase subunit C, mitochondrial {ECO:0000255; glutamyl-tRNA(Gln) amidotransferase, subunit C; glutamyl-tRNA(Gln) amidotransferase, subunit C homolog; HAMAP-Rule:MF_03149}; Protein 15E1.2; RGD1309698; similar to CG33084-PB, isoform B | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 283459 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.04 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O43716 | |
| GATC | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-136 of human GATC (NP_7897881). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction