missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GATC Polyclonal Antibody
GREENER_CHOICE

Product Code. 15902955
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15902955 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15902955 Supplier Invitrogen™ Supplier No. PA576426

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

GATC (glutamyl-tRNA(Gln) amidotransferase, subunit C) is a protein-coding gene. GO annotations related to this gene include glutaminyl-tRNA synthase (glutamine-hydrolyzing) activity. An important paralog of this gene is ENSG00000111780.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GATC
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene GATC
Gene Accession No. D3ZY68, O43716, Q8CBY0
Gene Alias 15E1.2; 2010003O18Rik; Gatc; gatC-like protein; glu-AdT subunit C; glu-AdT subunit C {ECO:0000255; glutamyl-tRNA amidotransferase subunit C; glutamyl-tRNA(Gln) amidotransferase subunit C, mitochondrial; glutamyl-tRNA(Gln) amidotransferase subunit C, mitochondrial {ECO:0000255; glutamyl-tRNA(Gln) amidotransferase, subunit C; glutamyl-tRNA(Gln) amidotransferase, subunit C homolog; HAMAP-Rule:MF_03149}; Protein 15E1.2; RGD1309698; similar to CG33084-PB, isoform B
Gene Symbols GATC
Host Species Rabbit
Immunogen Recombinant fusion protein of human GATC.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283459, 360821, 384281
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.