missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GATAD2B Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25679-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a zinc finger protein transcriptional repressor. The encoded protein is part of the methyl-CpG-binding protein-1 complex, which represses gene expression by deacetylating methylated nucleosomes. Mutations in this gene are linked to intellectual disability and dysmorphic features associated with cognitive disability.Specifications
| GATAD2B | |
| Polyclonal | |
| Unconjugated | |
| GATAD2B | |
| GATAD2B, KIAA1150, p66/p68, P66beta, RP11 216N14.6 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 57459 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8WXI9 | |
| GATAD2B | |
| GATAD2B Fusion Protein Ag22615 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction