missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GATAD2A Recombinant Rabbit Monoclonal Antibody (23GB5685), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | GATAD2A |
|---|---|
| Clone | 23GB5685 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089906
![]() |
Invitrogen™
MA553976 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088254
|
Invitrogen™
MA553977 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GATAD2A Recombinant Monoclonal antibody specifically detects GATAD2A in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| GATAD2A | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q86YP4, Q8CHY6 | |
| 234366, 54815 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB5685 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 1110066C11Rik; BC031407; C80248; GATA zinc finger domain containing 2A; GATA zinc finger domain-containing protein 2A; GATAD2A; Hp66alpha; MGC29315; MGC36975; p66 alpha; p66a; p66alpha; RGD1310596; transcriptional repressor p66 alpha; Transcriptional repressor p66-alpha | |
| A synthesized peptide derived from human p66 alpha (175-236AA). | |
| Primary | |
| -20°C | |
| GATAD2A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title