missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GATAD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568586
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 92%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 77%.
Component of some chromatin complex recruited to chromatin sites methylated 'Lys-4' of histone H3 (H3K4me).
Specifications
| GATAD1 | |
| Polyclonal | |
| Unconjugated | |
| GATAD1 | |
| 2310031E19Rik; 2810047M21Rik; 8430439A17Rik; 9130430G15Rik; B330017N08Rik; CMD2B; ENSMUSG00000073241; GATA zinc finger domain containing 1; GATA zinc finger domain-containing 1-like protein; GATA zinc finger domain-containing protein 1; GATA zinc finger domain-containing protein 1-like protein; Gatad1; im:7150682; ocular development associated; ocular development-associated gene protein; ODAG; RG083M05.2; RG83M5.2; si:dkey-231l1.7; Unknown (protein for MGC:136582); zgc:136582 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 57798 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8WUU5 | |
| GATAD1 | |
| synthetic peptide directed towards the middle region of human GATAD1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction