missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GATA6 Monoclonal Antibody (2F10G3)
GREENER_CHOICE

Product Code. 17450843 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17450843 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17450843 Supplier Invitrogen™ Supplier No. MA538529

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

This antibody has been tested in indirect ELISA.

The GATA6 gene encodes a member of the GATA family of zinc-finger transcription factors, which are critical in regulating gene expression during development. GATA6 plays a crucial role in the differentiation and development of multiple tissues, including the heart and gastrointestinal tract, by interacting with other transcriptional regulators through its zinc-finger domains. This gene influences signaling pathways that drive epithelial-to-mesenchymal transitions (EMTs) and immune surveillance, which are essential processes in maintaining tissue homeostasis and countering tumor progression. In the respiratory tract, GATA6 is essential for the differentiation of lung epithelial cells, impacting the expression of WNT7B and surfactant proteins, vital for proper lung function. Furthermore, studies suggest that GATA6 contributes to an immune-enriched phenotype linked to favorable prognostic outcomes in certain cancers, indicating its potential role in mitigating metastatic propensity by enhancing immune response and surveillance against tumorous cells.
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen GATA6
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 2F10G3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene GATA6
Gene Accession No. Q92908
Gene Alias AA410133; ASD9; AVSD5; DNA-binding protein GATA-GT2; GATA binding protein 6; Gata6; GATA-6; GATA-binding factor 6; GATA-binding protein 6; LOW QUALITY PROTEIN: transcription factor GATA-6; transcription factor GATA6; transcription factor GATA-6; zinc-finger transcription factor
Gene Symbols GATA6
Host Species Mouse
Immunogen Purified recombinant fragment of human GATA6 (aa: 491-557) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2627
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.