Learn More
Invitrogen™ GATA5 Monoclonal Antibody (1B11)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA515897
Description
MA5-15897 targets GATA5 in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15897 immunogen is purified recombinant fragment of human GATA5 expressed in E. Coli. . MA5-15897 detects GATA5 which has a predicted molecular weight of approximately 41kDa.
GATA binding factor 5 is a transcription regulator involved in embryonic development. In addition, the protein encoded by this gene contains two GATA-type zinc fingers and is known to bind to hepatocyte nuclear factor-1alpha (HNF-1alpha). This interaction is essential for cooperative activation of the intestinal lactase-phlorizin hydrolase promoter. GATA-5 may be involved in the establishment of cardiac smooth muscle cell diversity.
Specifications
| GATA5 | |
| Monoclonal | |
| Conc. Not Determined | |
| Ascites with 0.03% sodium azide | |
| Q9BWX5 | |
| Gata5 | |
| Purified recombinant fragment of human GATA5 expressed in E. Coli. | |
| RUO | |
| 140628 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Western Blot | |
| 1B11 | |
| Unconjugated | |
| Gata5 | |
| bB379O24.1; GATA binding factor-5; GATA binding protein 5; GATA5; GATA-binding factor 5; GATA-binding protein 5; GATAS; hypothetical protein LOC504784; transcription factor GATA-5; Transcription factor GATA-5 (GATA binding factor-5) | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.