missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GATA4 Recombinant Rat Monoclonal Antibody (eBioEvan), Alexa Fluor™ Plus 555

Rat Recombinant Monoclonal Antibody

Brand:  Invitrogen™ 740049TP55520UG

 View more versions of this product

Product Code. 30275693

  • £179.00 / 20µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Alexa Fluor™ Plus recombinant antibodies are conjugated using new, proprietary dye chemistry so you can generate stunning data. Alexa Fluor™ Plus antibodies represent an advancement in fluorescent conjugate technology. Alexa Fluor™ Plus antibodies provide brighter signal compared to leading Alexa Fluor™ antibodies, providing you with better signal-to-noise for your critical experiments. These antibodies show better specificity and lot-to-lot consistency as these are recombinant antibodies, generated by cloning specific genes for the desired antibodies into an expression vector and expressed in vitro. Using conjugate solutions: Centrifuge the protein conjugate solution briefly in a microcentrifuge before use; add only the supernatant to the experiment. This step will help eliminate any protein aggregates that may have formed during storage, thereby reducing nonspecific background staining. Applications Tested: This eBioEvan antibody has been tested by western blot, immunocytochemistry and flow cytometric analysis of Hep G2 cells. This may be used for immunocytochemistry at 5 μg/mL and for flow cytometry at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of inte...

GATA4 is a member of the GATA family of zinc-finger transcription factors. Members of this family recognize the GATA motif which is present in the promoters of many genes. This protein is thought to regulate genes involved in embryogenesis and in myocardial differentiation and function. Mutations in the gene encoding GATA4 have been associated with cardiac septal defects.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GATA4
Recombinant Monoclonal
1.0 mg/mL
Proprietary buffer with 0.008% Bromonitrodioxane, 0.008% Methylisothiazolone; pH 6.8
P43694, Q08369
GATA4
Affinity chromatography
RUO
14463, 2626
4°C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
eBioEvan
Alexa Fluor Plus 555
GATA4
ASD2; DNA-binding protein GATA-GT2; GAT4; GATA binding protein 4; GATA4; Gata-4; GATA-binding factor 4; GATA-binding protein 4; MGC126629; TACHD; TOF; Transcription factor GATA-4; VSD1
Rat
20 μL
Primary
Human, Mouse
Antibody
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.