missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GATA4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19530-1-AP-150UL
This item is not returnable.
View return policy
Description
The antibody is specific to GATA4.
This gene encodes a protein which contains a conserved nuclear localization signal. The encoded protein may have a role in spermatogenesis, cell cycle control, and in meiotic cell division.Specifications
| GATA4 | |
| Polyclonal | |
| Unconjugated | |
| GATA4 | |
| GATA binding factor 4, GATA binding protein 4, GATA4, Transcription factor GATA 4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14463, 2626, 54254 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.32 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P43694, P46152, Q08369 | |
| Gata4 | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido