missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GATA4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA523389
This item is not returnable.
View return policy
Description
GATA4 Polyclonal Antibody for Western Blot, IHC (P)
GATA4 is a member of the GATA family of zinc-finger transcription factors. Members of this family recognize the GATA motif which is present in the promoters of many genes. This protein is thought to regulate genes involved in embryogenesis and in myocardial differentiation and function. Mutations in the gene encoding GATA4 have been associated with cardiac septal defects.
Specifications
| GATA4 | |
| Polyclonal | |
| Unconjugated | |
| GATA4 | |
| ASD2; DNA-binding protein GATA-GT2; GAT4; GATA binding protein 4; GATA4; Gata-4; GATA-binding factor 4; GATA-binding protein 4; MGC126629; TACHD; TOF; Transcription factor GATA-4; VSD1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 14463, 2626 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| P43694, Q08369 | |
| GATA4 | |
| A portion of amino acids 330-380 of human GATA-4. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction