missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GATA4 Monoclonal Antibody (570CT10.5.7)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA537593
This item is not returnable.
View return policy
Description
GATA4 Monoclonal Antibody for Western Blot
GATA4 is a member of the GATA family of zinc-finger transcription factors. Members of this family recognize the GATA motif which is present in the promoters of many genes. This protein is thought to regulate genes involved in embryogenesis and in myocardial differentiation and function. Mutations in the gene encoding GATA4 have been associated with cardiac septal defects.
Specifications
| GATA4 | |
| Monoclonal | |
| Conc. Not Determined | |
| Ascites with 0.09% sodium azide; pH 7.4 | |
| P43694 | |
| GATA4 | |
| KLH conjugated synthetic peptide between 298-328 amino acids from the C-terminal region of human GATA4. | |
| RUO | |
| 2626 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 570CT10.5.7 | |
| Unconjugated | |
| GATA4 | |
| ASD2; DNA-binding protein GATA-GT2; GAT4; GATA binding protein 4; GATA4; Gata-4; GATA-binding factor 4; GATA-binding protein 4; MGC126629; TACHD; TOF; Transcription factor GATA-4; VSD1 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgM |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction