missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GATA4 Monoclonal Antibody (532025)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561187
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
GATA4 is a member of the GATA family of zinc-finger transcription factors. Members of this family recognize the GATA motif which is present in the promoters of many genes. This protein is thought to regulate genes involved in embryogenesis and in myocardial differentiation and function. Mutations in the gene encoding GATA4 have been associated with cardiac septal defects.
Specifications
| GATA4 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| P43694 | |
| GATA4 | |
| E. coli-derived recombinant human GATA-4, Met27-Ser212. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunocytochemistry | |
| 532025 | |
| Unconjugated | |
| GATA4 | |
| ASD2; DNA-binding protein GATA-GT2; GAT4; GATA binding protein 4; GATA4; Gata-4; GATA-binding factor 4; GATA-binding protein 4; MGC126629; TACHD; TOF; Transcription factor GATA-4; VSD1 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 2626 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction