missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GATA3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10417-1-AP-20UL
This item is not returnable.
View return policy
Description
This is a rabbit polyclonal antibody raised against part of C-terminus of human GATA3.
Trans-acting T-cell specific transcription factor GATA-3 is a member of GATA family of transcription factors that regulates development of multiple tissues. It is an important transcription factor in regulating human Th2 cell differentiation in vivo.Specifications
| GATA3 | |
| Polyclonal | |
| Unconjugated | |
| GATA3 | |
| GATA binding factor 3, GATA binding protein 3, GATA3, HDR | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14462, 2625, 85471 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.31 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P23771, P23772 | |
| Gata3 | |
| GATA3 Fusion Protein Ag0174 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction