Learn More
Invitrogen™ GATA3 Monoclonal Antibody (1A12-1D9)

Description
Western blot analysis of MA1-028 detects a prominent ~50 kDa protein in GATA3 expressing SH-SY5Y cells. The signal is enhanced in nuclear extracts due to enrichment of GATA3. This antibody has also been successfully tested in IP, IHC and Immunofluorescence applications. MA1-028 can be used for immunofluoresence analysis of Gata3 in human ESC lines. MA1-028 was successfully used in western blotting analysis of GATA3 in breast cancer cell lines.
Specifications
Specifications
| Antigen | GATA3 |
| Applications | ChIP Assay, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 1A12-1D9 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide |
| Gene | GATA3 |
| Gene Accession No. | P23771, P23772 |
| Gene Alias | EGK_19445; GATA binding factor-3; GATA binding protein 3; GATA binding protein-3; GATA3; Gata-3; GATA-binding factor 3; GATA-binding protein 3; HDR; HDRS; jal; MGC2346; MGC5199; MGC5445; Trans-acting T-cell-specific transcription factor GATA-3; Transacting T-cell-specific transcription factor GATA-3 |
| Show More |
Safety and Handling
- GATA3 Monoclonal Antibody (1A12-1D9)
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash hands thoroughly after handling.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contain: glycerol
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.