missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GATA1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA5143407
This item is not returnable.
View return policy
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
Transcriptional activator or repressor which probably serves as a general switch factor for erythroid development. It binds to DNA sites with the consensus sequence 5'-[AT]GATA[AG]-3' within regulatory regions of globin genes and of other genes expressed in erythroid cells.Specifications
GATA1 | |
Polyclonal | |
Unconjugated | |
eryf1; Erythroid transcription factor; erythroid transcription factor 1; GATA binding protein 1; GATA binding protein 1 (globin transcription factor 1); GATA1; GATA-1; GATA-binding factor 1; GATA-binding protein 1; GATA-binding protein 1 (globin transcription factor 1); GF1; Gf-1; globin transcription factor 1; HGNC:4170; NFE1; NF-E1; NF-E1 DNA-binding protein; nuclear factor, erythroid 1; transcription factor GATA1; XLANP; XLTDA; XLTT | |
Recombinant protein encompassing a sequence within the center region of zebrafish Gata1a. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (PFA fixed), Western Blot | |
1 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
Zebrafish | |
Antibody | |
IgG |