Learn More
Invitrogen™ GATA1 Monoclonal Antibody (4F5)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA515638
Description
MA5-15638 targets GATA1 in indirect ELISA, IF, IHC, and WB applications and shows reactivity with Human samples. The MA5-15638 immunogen is purified recombinant fragment of human GATA1 expressed in E. Coli. MA5-15638 detects GATA1 which has a predicted molecular weight of approximately 42.7.
Transcriptional activator or repressor which probably serves as a general switch factor for erythroid development. It binds to DNA sites with the consensus sequence 5'-[AT]GATA[AG]-3' within regulatory regions of globin genes and of other genes expressed in erythroid cells.
Specifications
| GATA1 | |
| Monoclonal | |
| Conc. Not Determined | |
| ascites with 0.03% sodium azide | |
| P15976 | |
| Gata1 | |
| Purified recombinant fragment of human GATA1 expressed in E. Coli. | |
| RUO | |
| 2623 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 4F5 | |
| Unconjugated | |
| Gata1 | |
| eryf1; Erythroid transcription factor; erythroid transcription factor 1; GATA binding protein 1; GATA binding protein 1 (globin transcription factor 1); GATA1; GATA-1; GATA-binding factor 1; GATA-binding protein 1; GATA-binding protein 1 (globin transcription factor 1); GF1; Gf-1; globin transcription factor 1; HGNC:4170; NFE1; NF-E1; NF-E1 DNA-binding protein; nuclear factor, erythroid 1; transcription factor GATA1; XLANP; XLTDA; XLTT | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.