missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GATA-4 Antibody (OTI9F9), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-47765
This item is not returnable.
View return policy
Description
GATA-4 Monoclonal specifically detects GATA-4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| GATA-4 | |
| Monoclonal | |
| 0.17 mg/mL | |
| Western Blot 1:200-500, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:50 | |
| GATA4 | |
| Human recombinant protein fragment corresponding to amino acids 296-442 of human GATA4 (NP_002043) produced in E.coli. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This antibody is specific for Homo sapiens GATA binding protein 4 (GATA4). | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| OTI9F9 | |
| Unconjugated | |
| GATA binding protein 4, GATA-binding factor 4, GATA-binding protein 4, MGC126629, transcription factor GATA-4 | |
| Mouse | |
| 44.4 kDa | |
| 0.1 mL | |
| Mesenchymal Stem Cell Markers, Stem Cell Markers | |
| 2626 | |
| Human, Mouse, Rat | |
| IgG3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction