missing translation for 'onlineSavingsMsg'
Learn More

GATA-4 Antibody (OTI9F9), mFluor Violet 450 SE, Novus Biologicals™

Product Code. 18412073 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18412073 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18412073 Supplier Novus Biologicals Supplier No. NBP270797MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

GATA-4 Monoclonal antibody specifically detects GATA-4 in Human, Mouse, Rat samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen GATA-4
Applications Flow Cytometry
Classification Monoclonal
Clone OTI9F9
Conjugate mFluor Violet 450 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias GATA binding protein 4, GATA-binding factor 4, GATA-binding protein 4, MGC126629, transcription factor GATA-4
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 296-442 of human GATA4 (NP_002043) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Mesenchymal Stem Cell Markers, Phospho Specific, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2626
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.