missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GATA-3 Mouse anti-Human, Clone: GATA3/2441, Novus Biologicals™
Mouse Monoclonal Antibody
£377.00
Specifications
| Antigen | GATA-3 |
|---|---|
| Clone | GATA3/2441 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1 to 2 μg/mL, Flow Cytometry 1 to 2 μg/million cells, ELISA, Protein Array |
| Applications | Western Blot, Flow Cytometry, ELISA, Peptide Array |
Description
GATA-3 Monoclonal specifically detects GATA-3 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Protein Array.Specifications
| GATA-3 | |
| 0.2 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Peptide Array | |
| Unconjugated | |
| Mouse | |
| Cancer, Innate Immunity | |
| 10 mM PBS with 0.05% BSA | |
| 2625 | |
| IgG2b κ | |
| Protein A or G purified |
| GATA3/2441 | |
| Western Blot 1 to 2 μg/mL, Flow Cytometry 1 to 2 μg/million cells, ELISA, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 131320, GATA binding protein 3, GATA-binding factor 3, GATA-binding protein 3, HDRMGC5199, MGC2346, MGC5445, trans-acting T-cell-specific transcription factor GATA-3 | |
| Recombinant fragment of human GATA-3 protein (around aa 357-436) (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title