missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
GATA-2 Monoclonal antibody specifically detects GATA-2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, KnockDown
Specifications
Specifications
| Antigen | GATA-2 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, KnockDown |
| Classification | Monoclonal |
| Clone | 2D11 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500, Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, Knockdown Validated |
| Formulation | In 1x PBS, pH 7.4 |
| Gene Accession No. | AAH18988 |
| Gene Alias | endothelial transcription factor GATA-2, FLJ45948, GATA binding protein 2, GATA-binding protein 2NFE1B, MGC2306 |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?