missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GATA-1 Antibody (4F5), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
£177.00
Specifications
| Antigen | GATA-1 |
|---|---|
| Clone | 4F5 |
| Dilution | Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, Chromatin Immunoprecipitation (ChIP) |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
GATA-1 Monoclonal specifically detects GATA-1 in Human, Baboon samples. It is validated for Western Blot, Chromatin Immunoprecipitation, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| GATA-1 | |
| Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, Chromatin Immunoprecipitation (ChIP) | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human | |
| 2623 | |
| Purified recombinant full length human GATA1 expressed in E. Coli. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| 4F5 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Stem Cell Markers | |
| Eryf1, ERYF1GATA-binding protein 1 (globin transcription factor 1), GATA binding protein 1 (globin transcription factor 1), GATA-1erythroid transcription factor, GATA-binding factor 1, GF-1, GF1globin transcription factor 1, NF-E1 DNA-binding protein, NFE1erythroid transcription factor 1, transcription factor GATA1, XLTT | |
| GATA1 | |
| IgG2b | |
| Unpurified | |
| 42.7 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title