missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAT3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£162.00 - £375.00
Spécification
| Antigen | GAT3 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
|
18331736
|
Novus Biologicals
NBP3-03494-20UL |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18341774
|
Novus Biologicals
NBP3-03494-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GAT3 Polyclonal specifically detects GAT3 in Human, Mouse, Rat samples. It is validated for Western Blot.Spécification
| GAT3 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| GABA Transporter 3, GABT3, GAT-3, GAT3sodium- and chloride-dependent GABA transporter 3, GAT4, solute carrier family 6 (neurotransmitter transporter, GABA), member 11, Solute carrier family 6 member 11 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-60 of human GAT3 (NP_055044.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Neuroscience | |
| PBS with 50% glycerol, pH7.3. | |
| 6538 | |
| IgG | |
| Affinity purified |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit