missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAT-1/SLC6A1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35498-20ul
This item is not returnable.
View return policy
Description
GAT-1/SLC6A1 Polyclonal antibody specifically detects GAT-1/SLC6A1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| GAT-1/SLC6A1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| GABA Transporter 1, GABATHG, GABATRGAT-1, GABT1, GAT1sodium- and chloride-dependent GABA transporter 1, solute carrier family 6 (neurotransmitter transporter, GABA), member 1, Solute carrier family 6 member 1 | |
| A synthetic peptide corresponding to a sequence within amino acids 500-599 of human GAT-1/SLC6A1 (NP_003033.3).,, Sequence:, WSFFTPIIVAGVFIFSAVQMTPLTMGNYVFPKWGQGVGWLMALSSMVLIPGYMAYMFLTLKGSLKQRIQVMVQPSEDIVRPENGPEQPQAGSSTSKEAYI | |
| 20 μL | |
| Neuroscience | |
| 6529 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction