missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GAT-1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA580031
This item is not returnable.
View return policy
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Rat Brain Tissue, Mouse Brain Tissue. IHC: Rat Cerebellum Tissue.
GAT-1 (SLC6A1) The protein encoded by SLC6A1 is a gamma-aminobutyric acid (GABA) transporter that localizes to the plasma membrane. The encoded protein removes GABA from the synaptic cleft, restoring it to presynaptic terminals.
Specifications
| GAT-1 | |
| Polyclonal | |
| Unconjugated | |
| SLC6A1 | |
| A730043E01; GABA T1; GABA T-1; GABA transporter 1; GABA transporter protein; GABAT1; GABATHG; GABATR; Gabt; GABT1; gamma-aminobutyric acid (GABA-A) transporter 1; GAT1; GAT-1; MAE; SLC6A1; Sodium- and chloride-dependent GABA transporter 1; solute carrier family 6 (neurotransmitter transporter), member 1; solute carrier family 6 (neurotransmitter transporter, GABA), member 1; solute carrier family 6 member 1; XT-1; Xtrp1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 232333, 6529, 79212 | |
| -20°C | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| 500 μg/mL | |
| PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal | |
| P23978, P30531, P31648 | |
| SLC6A1 | |
| A synthetic peptide corresponding to a sequence at the C-terminus of human GABA Transporter 1 (473-487aa WFYGVNRFYDNIQEM). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction