missing translation for 'onlineSavingsMsg'
Learn More

Gastrokine 1 Antibody [PerCP], Novus Biologicals™

Product Code. 30733226 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30733226 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30733226 Supplier Novus Biologicals Supplier No. AF7287C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Gastrokine 1 Polyclonal antibody specifically detects Gastrokine 1 in Human,Mouse samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Gastrokine 1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate PerCP
Formulation PBS
Gene Alias 18 kDa antrum mucosa protein, AMP-18, AMP18BRICHOS domain containing 1, BRICD1, CA11MGC70354, FOV, foveolin, gastrokine 1, gastrokine-1, Protein CA11
Host Species Sheep
Immunogen E. coli-derived recombinant mouse Gastrokine 1, Tyr38-Tyr201, Accession # Q9CR36
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 56287
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.