missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Gastrokine 1 Polyclonal antibody specifically detects Gastrokine 1 in Human samples. It is validated for ELISA
Specifications
Specifications
| Applications | ELISA |
| Classification | Polyclonal |
| Immunogen | Produced in rabbits immunized with purified, recombinant Human Gastrokine 1 (Uniprot#: Q9NS71; Asn35-Asn199) |
| Quantity | 0.1 mL |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?