missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Gastrokine 1 Polyclonal antibody specifically detects Gastrokine 1 in Human,Mouse samples. It is validated for Immunohistochemistry,Western Blot
Specifications
Specifications
| Antigen | Gastrokine 1 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | mFluor Violet 500 SE |
| Formulation | 50mM Sodium Borate |
| Gene Alias | 18 kDa antrum mucosa protein, AMP-18, AMP18BRICHOS domain containing 1, BRICD1, CA11MGC70354, FOV, foveolin, gastrokine 1, gastrokine-1, Protein CA11 |
| Host Species | Sheep |
| Immunogen | E. coli-derived recombinant mouse Gastrokine 1, Tyr38-Tyr201, Accession # Q9CR36 |
| Purification Method | Antigen Affinity-purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?