missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gastrokine 1 Antibody (2E5), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00056287-M01
This item is not returnable.
View return policy
Description
Gastrokine 1 Monoclonal antibody specifically detects Gastrokine 1 in Human samples. It is validated for Western Blot, ELISA, ELISA
Specifications
| Gastrokine 1 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| 18 kDa antrum mucosa protein, AMP-18, AMP18BRICHOS domain containing 1, BRICD1, CA11MGC70354, FOV, foveolin, gastrokine 1, gastrokine-1, Protein CA11 | |
| GKN1 (AAH59778, 21 a.a. ~ 185 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. NYNINVNDDNNNAGSGQQSVSVNNEHNVANVDNNNGWDSWNSIWDYGNGFAATRLFQKKTCIVHKMNKEVMPSIQSLDALVKEKKLQGKGPGGPPPKGLMYSVNPNKVDDLSKFGKNIANMCRGIPTYMAEEMQEASLFFYSGTCYTTSVLWIVDISFCGDTVEN | |
| 0.1 mg | |
| Cell Cycle and Replication | |
| 56287 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, ELISA, Sandwich ELISA | |
| 2E5 | |
| Western Blot 1:500, ELISA, Sandwich ELISA | |
| AAH59778 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction