missing translation for 'onlineSavingsMsg'
Learn More

Gastrodin, MedChemExpress

Product Code. 16468986
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16468986 100 mg 100mg
16463836 25 mg 25mg
16454757 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16468986 Supplier Medchem Express Supplier No. HYN0115
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Gastrodin, a main constituent of a Chinese herbal medicine Tianma, has been known to display anti-inflammatory effects. Gastrodin, has long been used for treating dizziness, epilepsy, stroke and dementia.

Gastrodin, a main constituent of a Chinese herbal medicine Tianma, has been known to display anti-inflammatory effects. Gastrodin, has long been used for treating dizziness, epilepsy, stroke and dementia.

Specifications

Chemical Name or Material Gastrodin
CAS 62499-27-8
Color White
Purity Grade Notes Research
Molecular Formula C13H18O7
Quantity 100 mg
Synonym Gastrodine
Solubility Information DMSO : ≥ 100 mg/mL (349.31 mM) ∣H2O : 100 mg/mL (349.31 mM; Need ultrasonic)
SMILES O[C@H]([C@H]([C@@H]([C@@H](CO)O1)O)O)[C@@H]1OC2=CC=C(CO)C=C2
Molecular Weight (g/mol) 286.28
Formula Weight 286.28
Percent Purity 99.14%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.