missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Gastrin-releasing Peptide R/GRPR Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£164.00 - £442.00
Tekniske data
| Antigen | Gastrin-releasing Peptide R/GRPR |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Produktkode | Brand | Quantity | Pris | Antal & tilgængelighed | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkode | Brand | Quantity | Pris | Antal & tilgængelighed | |||||
|
30227852
|
Novus Biologicals
NBP3-38023-100ul |
100 μL |
£442.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30231228
|
Novus Biologicals
NBP3-38023-20ul |
20 μL |
£164.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beskrivelse
Gastrin-releasing Peptide R/GRPR Polyclonal antibody specifically detects Gastrin-releasing Peptide R/GRPR in Human,Mouse,Rat samples. It is validated for ELISA,Western BlotTekniske data
| Gastrin-releasing Peptide R/GRPR | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| GPCR | |
| PBS (pH 7.3), 50% glycerol | |
| 2925 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| gastrin-releasing peptide receptor, GRP-preferring bombesin receptor, GRP-R | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 235-384 of human Gastrin-releasing Peptide R/GRPR (NP_005305.1).,, Sequence:, KNLIQSAYNLPVEGNIHVKKQIESRKRLAKTVLVFVGLFAFCWLPNHVIYLYRSYHYSEVDTSMLHFVTSICARLLAFTNSCVNPFALYLLSKSFRKQFNTQLLCCQPGLIIRSHSTGRSTTCMTSLKSTNPSVATFSLINGNICHERYV | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel