missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gastrin-releasing Peptide/Bombesin/Neuromedin C Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£171.00 - £386.00
Specifications
| Antigen | Gastrin-releasing Peptide/Bombesin/Neuromedin C |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18330997
|
Novus Biologicals
NBP3-03282-20UL |
20 μL |
£171.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18043486
|
Novus Biologicals
NBP3-03282-100UL |
100 μL |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Gastrin-releasing Peptide/Bombesin/Neuromedin C Polyclonal specifically detects Gastrin-releasing Peptide/Bombesin/Neuromedin C in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| Gastrin-releasing Peptide/Bombesin/Neuromedin C | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| BN, bombesin, gastrin-releasing peptide, GRP-10, neuromedin C, pre-progastrin releasing peptide, preproGRP, proGRP | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-148 of human Gastrin-releasing Peptide/Bombesin/Neuromedin C (NP_002082.2). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| GPCR, Neuroscience | |
| PBS with 50% glycerol, pH7.3. | |
| 2922 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title