missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Gastrin Recombinant Rabbit Monoclonal Antibody (PD00-37)
GREENER_CHOICE

Product Code. 17858072
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17858072 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17858072 Supplier Invitrogen™ Supplier No. MA544651

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Gastrin Recombinant Monoclonal Antibody for IHC (P)

This antibody may be used to study the differential tissue expression and intracellular and subcellular localization of gastrin in neuroendocrine cells of the gastrointestinal tract, and in the CNS. Antibodies to gastrin are also useful for the identification and detection of gastrin in normal and neoplastic tissue.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Gastrin
Applications Immunohistochemistry (Paraffin)
Classification Recombinant Monoclonal
Clone PD00-37
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol, 0.1% BSA and 0.05% sodium azide; pH 7.4
Gene Gast
Gene Accession No. P01350
Gene Alias Big gastrin; G14; G17; G34; G52; G6; G71; GAS; Gast; Gastrin; Gastrin component I; Gastrin component II; Gastrin component III; gastrin-1; Gastrin-14; Gastrin-17; Gastrin-34; Gastrin-52; Gastrin-6; Gastrin-71; PPG34; preprogastrin
Gene Symbols Gast
Host Species Rabbit
Immunogen Synthetic peptide within Human Gastrin (internal sequence).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2520
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.